HPLC columns, Hypersil GOLD™ C18
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25005-102130 25005-154030 25005-153030 25005-152130 25005-054630 25005-053030 25005-052130 25005-014001 25005-013001 25005-104630 25005-154630 25005-254630 25005-253030 25003-012101 25003-011001 25003-014001 25005-012101 25003-103030 25003-154030 25003-153030 25003-152130 25003-124630 25003-013001 25003-104030 25003-102130 25003-101030 25003-054630 25003-053030 25003-052130 25003-034630 25003-032130 25003-104630 25003-154630 25005-104030 25005-011001 25003-051030 25003-254030
554-4295EA 7530 DKK
554-4295 554-4380 554-4567 554-4297 554-4294 554-4561 554-4293 554-4557 554-4556 554-4296 554-4298 554-4299 554-4570 554-4530 554-4529 554-4532 554-4555 554-4546 554-4552 554-4551 554-4291 554-4549 554-4531 554-4547 554-4289 554-4545 554-4288 554-4541 554-4287 554-4539 554-4537 554-4290 554-4292 554-4565 554-4554 554-4540 554-4553
HPLC columns, Hypersil GOLD™ C18
Kromatografikolonner
Endcapped, ultra-pure, silica-based columns with exceptional peak shape and resolution designed for HPLC and  LC/MS.
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitivity
  • Confidence in the accuracy and quality of analytical data
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in theMore Product Information
Bestil nu
Specifikationer
Environmentally Preferable
Kulstofbelastning 10%
pH range 1 - 11
Surface area 220 m²/g