UHPLC columns, Hypersil GOLD™ aQ
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25302-152130 25302-202130 25302-102130 25302-052130 25302-051030 25302-103030 25302-101030 25302-054630
554-4386EA 8143 DKK
554-4386 554-4387 554-4384 554-4307 554-4648 554-4385 554-4649 554-4383
UHPLC columns, Hypersil GOLD™ aQ
Kromatografikolonner
Hypersil GOLD™ aQ polar endcapped C18 columns provide a controlled interaction mechanism by which polar analytes can be retained and resolved.
  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Excellent peak shape
  • Stable in 100% aqueous mobile phase
The additional interaction mechanism often provides selectivity differences over the traditional alkyl chain chemistries, and offers a solution for the separation of polar compounds which exhibit insufficient retention on pure alkyl chain phases under typical reversed phase mobile phase conditions.