Detergents for Glassware Washers
Leverandør: SMEG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DETERLQC2 ACIDGLP2 ACIDGLC2 DETERLQSP2 DETERLIQUIDI2
SMEGDETERLQC2EA 873 DKK
SMEGDETERLQC2 SMEGACIDGLP2 SMEGACIDGLC2 SMEGDETERLQSP2 SMEGDETERLIQUIDI2
Detergents for Glassware Washers
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Detergents designed to optimise cleaning efficiency and the thermal disinfection phase for laboratory glassware cleaning.
  • For thorough instrument cleaning
  • Alkaline detergents for washing and neutralisers for the neutralisation stage
DETERGLASS
• Universal alkaline powder
• Detergent for automatic washing of laboratory glassware
• Contains sodium hydroxide, sodium metasilicate pentahydrate, sodium triphosphate pentabasic, triclosene sodium dihydrate
• 10 kg pack
DETERLIQUID C2
• Strong universal liquid alkaline detergent phosphate-free for washing steel up to 95 °C
• Suitable for washing rubber and plastic at 60 °C
• Containing sodium silicate, polyacrylate, potassium hydroxide
• 5 litre can
ACIDGLASS C2
• Liquid phosphate-free neutraliser
• Containing citric acid monohydrate
• 5 litre can
ACIDGLASS P2
• Acid liquid neutralising agent for laboratory glassware
• Containg phosphoric acid, citric acid monohydrate
• 5 litre can