ESD occupational shoes, clogs, HACCP, Dynamic, 37360
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37360/39 37360/35 37360/45 37360/44 37360/43 37360/42 37360/41 37360/40 37360/38 37360/36 37360/37 37360/48 37360/46 37360/47
ABEB37360/39EA 446 DKK
ABEB37360/39 ABEB37360/35 ABEB37360/45 ABEB37360/44 ABEB37360/43 ABEB37360/42 ABEB37360/41 ABEB37360/40 ABEB37360/38 ABEB37360/36 ABEB37360/37 ABEB37360/48 ABEB37360/46 ABEB37360/47
ESD occupational shoes, clogs, HACCP, Dynamic, 37360
Sko og støvler
ESD clog made from smooth microfibre with a non foldable, adjustable heel strap. The TPU sole is resistant to fuel, oil and animal fats.
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
  • Washable at 30 °C
Colour: White
Certificeringer: EN 61340 (ESD); HACCP; DGUV 112-191; EN ISO 20347:2012; OB, FO,E,A, SRC.