ESD occupational shoes, clogs, Dynamic, 37343
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37343/44 37343/36 37343/35 37343/38 37343/39 37343/37 37343/48 37343/47 37343/46 37343/45 37343/43 37343/42 37343/41 37343/40
ABEB37343/44EA 465 DKK
ABEB37343/44 ABEB37343/36 ABEB37343/35 ABEB37343/38 ABEB37343/39 ABEB37343/37 ABEB37343/48 ABEB37343/47 ABEB37343/46 ABEB37343/45 ABEB37343/43 ABEB37343/42 ABEB37343/41 ABEB37343/40
ESD occupational shoes, clogs, Dynamic, 37343
Sko og støvler
ESD clog made from smooth microfibre with textile inlays and a non foldable, adjustable heel strap. The TPU sole is resistant to fuel, oil and animal fats.
  • Perspiration-absorbent lining
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
  • Washable at 30 °C
Colour: Black/yellow
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; OB, A, E, FO, SRC