Insoles for safety shoes, 3553, static control, acc Wave
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3553/36 3553/37 3553/38 3553/39 3553/40 3553/41 3553/42 3553/43 3553/44 3553/45 3553/46 3553/47
ABEB3553/36EA 121 DKK
ABEB3553/36 ABEB3553/37 ABEB3553/38 ABEB3553/39 ABEB3553/40 ABEB3553/41 ABEB3553/42 ABEB3553/43 ABEB3553/44 ABEB3553/45 ABEB3553/46 ABEB3553/47
Insoles for safety shoes, 3553, static control, acc Wave
Sko indlægssåler
Anti-static acc Wave (air clima comfort) insoles made from breathable fabric.
  • Beneficial effect for joints, musculature, ligaments and spine cushioning over the whole surface
  • Pressure absorbing, climatising and comfortable
Colour: White/blue
Certificeringer: EN 61340 (ESD).