Cleanroom wipes, pre-saturated, sterile, Sterile TechniSat®
Leverandør: Texwipe
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TX3217 TX3214
115-0314EA 6370 DKK
115-0314 115-1777
Cleanroom wipes, pre-saturated, sterile, Sterile TechniSat®
Papirservietter Servietter til Renrum Imprægneret
These wipes constructed from a hydroentangled polyester/cellulose blend are pre-wetted with 70% isopropyl alcohol and 30% DIW (filtered through a 0,2 μm filter). Each package is gamma irradiated to 10⁻⁶ Sterility Assurance Level.
  • Binder-free, hydroentangled polyester/cellulose substrate
  • Packaging permits alcohol wipedown of exterior bag before introduction into sterile suites
  • Offers low levels of solvent extractables and particle generation
  • Ideal for wiping gloved hands
Ideal for alcohol wiping in sterile environments in pharmaceutical aseptic fill areas, biotechnology manufacturing facilities, microbiological laboratories, sterile suites and prep rooms.
Emballering: Packaged in a resealable FlexPack.