ESD safety ankle boots, Velcro® fastening, HACCP, Anatom, 32280
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32280/39 32280/36 32280/47 32280/48 32280/45 32280/44 32280/41 32280/40
ABEB32280/39EA 870 DKK
ABEB32280/39 ABEB32280/36 ABEB32280/47 ABEB32280/48 ABEB32280/45 ABEB32280/44 ABEB32280/41 ABEB32280/40
ESD safety ankle boots, Velcro® fastening, HACCP, Anatom, 32280
Sko og støvler
ESD ankle boot made from microfibre, reflective material, Velcro® fastening, cushioned ankle support and a fuel resistant PU outsole with high slip resistance and self-cleaning profile.
  • Cold insulating lining
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for use in kitchens.
Colour: White
Certificeringer: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S3, SRC.