ESD occupational shoes, slip-on, X-Light, 7131132/7131137
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7131132/35 7131132/36 7131132/37 7131132/38 7131132/39 7131132/40 7131132/41 7131132/42 7131132/43 7131132/44 7131132/45 7131132/46 7131132/47 7131132/48 7131137/35 7131137/36 7131137/37 7131137/38 7131137/39 7131137/40 7131137/41 7131137/42 7131137/43 7131137/44 7131137/45 7131137/46 7131137/47 7131137/48
ABEB7131132/35EA 610 DKK
ABEB7131132/35 ABEB7131132/36 ABEB7131132/37 ABEB7131132/38 ABEB7131132/39 ABEB7131132/40 ABEB7131132/41 ABEB7131132/42 ABEB7131132/43 ABEB7131132/44 ABEB7131132/45 ABEB7131132/46 ABEB7131132/47 ABEB7131132/48 ABEB7131137/35 ABEB7131137/36 ABEB7131137/37 ABEB7131137/38 ABEB7131137/39 ABEB7131137/40 ABEB7131137/41 ABEB7131137/42 ABEB7131137/43 ABEB7131137/44 ABEB7131137/45 ABEB7131137/46 ABEB7131137/47 ABEB7131137/48
ESD occupational shoes, slip-on, X-Light, 7131132/7131137
Sko og støvler
ESD shoe made from smooth leather upper and lightweight PU walking sole, resistant to hydrocarbons.
  • Perspiration-absorbent special lining with silver ions
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Instep area with elastic band
Colour: White
Colour: White or black
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O2, FO, SRC.