ESD safety shoes, clogs, X-Light, 7131041/7131042
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7131041/35 7131041/36 7131041/37 7131041/38 7131041/39 7131041/40 7131041/41 7131041/42 7131041/43 7131041/44 7131041/45 7131041/46 7131041/47 7131041/48 7131042/35 7131042/36 7131042/37 7131042/38 7131042/39 7131042/40 7131042/41 7131042/42 7131042/43 7131042/44 7131042/45 7131042/46 7131042/47 7131042/48
ABEB7131041/35EA 616 DKK
ABEB7131041/35 ABEB7131041/36 ABEB7131041/37 ABEB7131041/38 ABEB7131041/39 ABEB7131041/40 ABEB7131041/41 ABEB7131041/42 ABEB7131041/43 ABEB7131041/44 ABEB7131041/45 ABEB7131041/46 ABEB7131041/47 ABEB7131041/48 ABEB7131042/35 ABEB7131042/36 ABEB7131042/37 ABEB7131042/38 ABEB7131042/39 ABEB7131042/40 ABEB7131042/41 ABEB7131042/42 ABEB7131042/43 ABEB7131042/44 ABEB7131042/45 ABEB7131042/46 ABEB7131042/47 ABEB7131042/48
ESD safety shoes, clogs, X-Light, 7131041/7131042
Sko og støvler
ESD safety shoe made from smooth leather and a lightweight PU sole (fuel resistant).
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Foldable, adjustable heel strap
  • Instep area with elastic band
Colour: White or black
Colour: White/black
Certificeringer: EN 61340 (ESD); DGUV 112-191; SB, A, E, FO, SRC.