Mailing boxes
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95.900 95.901 95.902
SARS95.900EA 128.53 DKK
SARS95.900 SARS95.901 SARS95.902
Mailing boxes
Transportbeholdere
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
  • With printed label
  • Lightweight
  • Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.