Stirrer shafts, double-moon shape, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C374-12 C374-18 C374-14
BOHLC374-12EA 2730 DKK
BOHLC374-12 BOHLC374-18 BOHLC374-14
Stirrer shafts, double-moon shape, BOLA
Omrørere Omrøreaksler
Stirrer shafts made of PTFE-jacketed stainless steel, two each tilting half-moon stirrer blades with double-sided groove and access for the stirrer shaft completely made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Creates tangential flow with little turbulence
  • With chucking diameter of 8 mm
Half-moon blade is ideal for stirring in round-bottom flasks with ground joint neck.