ESD safety shoes, sandals, HACCP, Velcro® fastening, Anatom, 2616
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2616/41 2616/42 2616/45 2616/36 2616/37 2616/38 2616/39 2616/40 2616/43 2616/44 2616/46 2616/47 2616/48 2616/49 2616/50
114-1155EA 725 DKK
114-1155 114-1156 114-1159 114-1150 114-1151 114-1152 114-1153 114-1154 114-1157 114-1158 114-1160 114-1161 114-1162 114-7409 114-7410
ESD safety shoes, sandals, HACCP, Velcro® fastening, Anatom, 2616
Sko og støvler
ESD safety shoe made from microfibre upper, reflective material, Velcro® fastening and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration absorbing special lining with silver fibre
  • Replaceable acc Wave insole, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and movement of the foot
  • TPU torsion waist, arch support and high stability
  • Shock absorbent PU midsole
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for hygienically demanding areas.
Colour: White
Certificeringer: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S1, SRC.