ESD safety shoes, clogs, Classic, 31000
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31000/36 31000/37 31000/38 31000/39 31000/40 31000/41 31000/42 31000/43 31000/44 31000/45 31000/46
ABEB31000/36EA 531 DKK
ABEB31000/36 ABEB31000/37 ABEB31000/38 ABEB31000/39 ABEB31000/40 ABEB31000/41 ABEB31000/42 ABEB31000/43 ABEB31000/44 ABEB31000/45 ABEB31000/46
ESD safety shoes, clogs, Classic, 31000
Sko og støvler
ESD clog made from grained leather, a non foldable, adjustable heel strap and PU sole (resistant to animal fats, oil and fuel).
  • Comfortable shape
  • Leather lining, heel patch is microfibre
  • Slip resistant PU sole
  • Protective steel toecap (200 J)
  • Adjustable instep strap with buckle fastening
Colour: White
Certificeringer: EN 61340 (ESD); EN ISO 20345:2011; SB, A, E, FO, WRU, SRC.