VWR® Q Path® MacroStar I, Cassettes, in Dispenser
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2373EA 2840 DKK
720-2373 720-2374 720-2375 720-2376 720-2377 720-2378 720-2379 720-2380 720-2381
VWR® Q Path® MacroStar I, Cassettes, in Dispenser
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Suitable for stainless steel lids. Resistant to all commonly used histological solvents. Cassettes with tape (= rack) for Leica and Sakura cassettes printers. Rack with 40 cassettes.
  • Large opening for the filtration of fluids
  • Without lid, with 34 holes
  • Designed to accept standard metal lids (M481)
  • Slanted writing surface (45° angle) accepts markings easily
  • Colour coded