Micro protein quantification set, QuantiMate
Leverandør: BIOCHROM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80-7100-70 80-7100-71 80-7100-72 80-7100-80 80-7100-81 80-7100-82
730-1450EA 0 DKK
730-1450 730-1451 730-1452 730-1453 730-1454 730-1455
Micro protein quantification set, QuantiMate
Kuvetter Spektrofotometerkuvetter
QuantiMate is designed for concentration and purity determinations of nucleic acids and proteins.
  • Measurement performance giving accurate and reproducible concentration results
  • Samples measured without time consuming dilutions
  • Only one cuvette is required
Dimensions are equivalent to standard cuvettes in order to work in most spectrophotometers.