GC septa, TR-Blue
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
313B3233 313B3240
555-8258EA 396 DKK
555-8258 555-8259
GC septa, TR-Blue
Septa
Made of low-bleed silicone, these septa have excellent mechanical properties and are ideal for demanding GC-MS applications.
  • General purpose septa
  • Easy to penetrate
  • Maximum temperature 200 to 250 °C