VWR® Q Path® Quickspot, Slide Holders for Turbitec®
Leverandør: LABONORD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05305194
630-2095EA 2860 DKK
630-2095 630-2094 630-2093
VWR® Q Path® Quickspot, Slide Holders for Turbitec®
Objektglas holdere
Q Path® Quickspot and slide holder for Turbitec are used to carry out cytocentrifugation. They allow a cell suspension to be deposited on a slide (gynaecological specimens). The slide holder is a support for positioning the slide and Quickspot is the sample filling chamber.
  • Q Path® slide holder (reusable) and Q Path® Quickspot (single use)
  • Accessories fit together and are used together
  • Resistant to centrifugation up to 2039×g for 30 min
Please note: For use with Hettich® centrifuges (Rotofix, Rotina) and compatible rotor and buckets.