Funnels for Tharmac® cytocentrifuge
Leverandør: THARMAC GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JC1395-200 JC1391-40
720-1795EA 798 DKK
720-1795 720-2676
Funnels for Tharmac® cytocentrifuge
Tragte Cytologitragte
Suitable only with EASY rotors JC105 and JC375.
  • EASY double cytology funnel with white filter card
  • Disposable sample chambers with disposable clip
  • For sample volumes of up to 0,5 ml
  • Two sedimentation fields with a diameter of 6mm each
  • Ideal for immunocytochemical processes
  • EASY ECOfunnel® with ECOseal
  • Disposable sample chamber with disposable clip
  • For preparing large-volume samples of up to 6ml
  • Sedimentation field of 22×15 mm for a higher cell yield
  • ECOträger slides (with JC1391/25 included)