IP Biopsy VI cassettes, taped
Leverandør: LEICA HISTOLOGY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39LC-550-1-L 39LC-550-10-L 39LC-550-11-L 39LC-550-2-L 39LC-550-3-L 39LC-550-4-L 39LC-550-6-L 39LC-550-7-L 39LC-550-8-L 39LC-550-9-L 39LC-550-5-L
720-2280EA 1010 DKK
720-2280 720-2472 720-2474 720-2476 720-2478 720-2480 720-2484 720-2486 720-2488 720-2490 720-2482
IP Biopsy VI cassettes, taped
Histologikassetter Indstøbningskassetter, Biopsiprøver
IP Biopsy VI cassettes are one piece cassettes with frangible hinges for easy lid removal, a 45° writing surface and a secure latch closure. The cassettes have been designed for optimal workflow in the Leica IP C Cassette Printer and offer <1% printing error rate.
  • Available in taped packaging for easy printer loading
  • Eleven colours in quantities of 1000 available