HPLC columns, Fluophase
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
82705-052130 82705-254630 82705-153030
HYPE82705-052130EA 8510 DKK
HYPE82705-052130 HYPE82705-254630 HYPE82705-153030
HPLC columns, Fluophase
Kromatografikolonner
Columns with perfluorinated phases provide unique selectivity.
  • Extra retention and selectivity for halogenated compounds
  • Ideal for taxane analysis