ESD cleanroom shoe covers, 3900/3910
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3900/5 3910/2 3910/3 3900/1 3900/3 3910/5 3910/4
ABEB3900/5EA 387 DKK
ABEB3900/5 ABEB3910/2 ABEB3910/3 ABEB3900/1 ABEB3900/3 ABEB3910/5 ABEB3910/4
ESD cleanroom shoe covers, 3900/3910
Arbejdstøj
The upper material of these shoe covers is Integrity® 1750 fabric. This densely-woven, polyester filament, grid fabric is manufactured with a 6-µ pore size, which filters out critical >0,5 µ particles with greater efficiency than typical Class 100 (ISO Class 5) fabrics used today.
  • Non migrating antimicrobial agent inhibits the growth of a wide range of problem-causing bacteria, mould, mildew and fungi
  • Carbon grid woven into the fabric controls electro-inductive migration
  • High moisture vapor transport assures wearer comfort
Suitable for use in Class 100, ISO 5, M3.5 cleanrooms.
Colour: White or light blue