ESD occupational shoes, clogs, Arrow, 38600/38602
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
38600/36 38600/37 38600/38 38600/39 38600/40 38600/41 38600/42 38600/43 38600/44 38600/45 38600/46 38600/47
ABEB38600/36EA 560 DKK
ABEB38600/36 ABEB38600/37 ABEB38600/38 ABEB38600/39 ABEB38600/40 ABEB38600/41 ABEB38600/42 ABEB38600/43 ABEB38600/44 ABEB38600/45 ABEB38600/46 ABEB38600/47
ESD occupational shoes, clogs, Arrow, 38600/38602
Sko og støvler
ESD clogs made from perforated leather, leather lining and a non foldable but adjustable heel strap. See model 38200.
  • Padded instep area with elastic band and Velcro® fastening at the heel
  • Sanitized® treatment reduces odour and bacterial growth
  • Abrasion and slip resistant PU sole
Colour: White
Certificeringer: EN 61340 (ESD); EN ISO 20347:2012; OB, A, FO, SRA.