Insoles for occupational shoes, 3565/3566, Reflexor
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3565/36 3565/37 3565/38 3565/39 3565/40 3565/41 3565/42 3566/40 3566/41 3566/42 3566/43 3566/44 3566/45 3566/46
ABEB3565/36EA 124 DKK
ABEB3565/36 ABEB3565/37 ABEB3565/38 ABEB3565/39 ABEB3565/40 ABEB3565/41 ABEB3565/42 ABEB3566/40 ABEB3566/41 ABEB3566/42 ABEB3566/43 ABEB3566/44 ABEB3566/45 ABEB3566/46
Insoles for occupational shoes, 3565/3566, Reflexor
Sko indlægssåler
Anti-static insoles made from microfibre.
  • Reflexor massage active-integrated insole
  • Comfortable orthopaedic integrated insole
  • Massage knobs stimulate blood circulation
Colour: Anthracite.
Colour: Anthracite
Certificeringer: EN 61340 (ESD)