General purpose gloves, Perfect Poly® PU First Grey
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2100250/10 2100250/11 2100250/9 2100250/6 2100250/7 2100250/8
DALL2100250/10EA 232 DKK
DALL2100250/10 DALL2100250/11 DALL2100250/9 DALL2100250/6 DALL2100250/7 DALL2100250/8
General purpose gloves, Perfect Poly® PU First Grey
Handsker
100% polyamide knitted glove with grey PU coating on palm and fingers.
  • Ventilated back optimise the glove's breathability
  • PU coating ensures a good grip at the fingertips
  • Polyamide and PU coating provide excellent abrasion and tear resistance
  • Gloves are certified as 'silicone-free' to ensure their use in painting environment
  • Non pilling, the polyamide is suitable for precision work
Ideal for the handling of fine objects in a dry environment or slightly greasy or dirty objects.
Certificeringer: EN 388:2016 (3131F), Cat. II