ESD occupational shoes, clogs, Arrow, 38200/38210
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
38200/36 38200/37 38200/38 38200/39 38200/40 38200/41 38200/42 38200/43 38200/44 38200/45 38200/46 38200/47 38210/36 38210/37 38210/38 38210/39 38210/40 38210/41 38210/42 38210/43 38210/44 38210/45 38210/46 38210/47
ABEB38200/36EA 478 DKK
ABEB38200/36 ABEB38200/37 ABEB38200/38 ABEB38200/39 ABEB38200/40 ABEB38200/41 ABEB38200/42 ABEB38200/43 ABEB38200/44 ABEB38200/45 ABEB38200/46 ABEB38200/47 ABEB38210/36 ABEB38210/37 ABEB38210/38 ABEB38210/39 ABEB38210/40 ABEB38210/41 ABEB38210/42 ABEB38210/43 ABEB38210/44 ABEB38210/45 ABEB38210/46 ABEB38210/47
ESD occupational shoes, clogs, Arrow, 38200/38210
Sko og støvler
ESD clogs made from smooth leather with leather lining, microfibre heel patch and a foldable and adjustable heel strap.
  • Abrasion and slip resistant PU sole
  • Sanitized® treatment reduces odour and bacterial growth
Colour: White or black
Colour: White/black
Certificeringer: EN 61340 (ESD); EN ISO 20347:2012; OB, A, FO, SRA.