Clamps for standard ground joints
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5836500 5836502 5836503 5836504 5836505 5836506 5836507
MARI5836500EA 11.5 DKK
MARI5836500 MARI5836502 MARI5836503 MARI5836504 MARI5836505 MARI5836506 MARI5836507
Clamps for standard ground joints
Klemmer Klemmer til NS-slib
These clamps made of POM for standard ground joints are used to securely link the socket and the cone.
  • Excellent chemical resistance
  • Material avoids scratching on the glass surfaces
  • Can be used at temperatures up to +150 °C
  • Clips are colour coded for easy identification