ESD occupational shoes, lace-up, Uni6, 36721
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36721/38 36721/35 36721/36 36721/37 36721/39 36721/40 36721/41 36721/42 36721/43 36721/44 36721/45 36721/46 36721/47 36721/48
ABEB36721/38EA 765 DKK
ABEB36721/38 ABEB36721/35 ABEB36721/36 ABEB36721/37 ABEB36721/39 ABEB36721/40 ABEB36721/41 ABEB36721/42 ABEB36721/43 ABEB36721/44 ABEB36721/45 ABEB36721/46 ABEB36721/47 ABEB36721/48
ESD occupational shoes, lace-up, Uni6, 36721
Sko og støvler
ESD shoes made from suede with breathable textile inlays, reflective material and a TPU sole resistant to oils, fuels and animal fats.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • PU midsole
  • Slip resistant TPU sole
Colour: Black
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O1, FO, SRC.