High pressure tees and crosses, PEEK
Leverandør: VICI JOUR RESEARCH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JR-1030 JR-1040 JR-2030 JR-2040 JR-1043 JR-1042 JR-1033 JR-1032 JR-2043 JR-2033 JR-2032 JR-2042
554-1023EA 2600 DKK
554-1023 554-1024 JOURJR-2030 JOURJR-2040 JOURJR-1043 JOURJR-1042 JOURJR-1033 JOURJR-1032 JOURJR-2043 JOURJR-2033 JOURJR-2032 JOURJR-2042
High pressure tees and crosses, PEEK
Fittings og stik
PEEK tees and crosses, high pressure, for easy connection of solvent lines.
  • Low dead volume
  • Completely inert and biocompatible