Chemical resistant gloves, Tricopren® 723
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
723.8 723.9 723.10 723.11
112-1751EA 470 DKK
112-1751 112-1752 112-1753 112-1754
Chemical resistant gloves, Tricopren® 723
Handsker
This glove is dipped to form the chloroprene layer, then it is dipped in a solvent-free adhesive bath and after that the cotton tricot is pulled on. Thus, the glove is fully coated, impermeable and provides a high chemical and mechanical resistance.
  • Free from latex
  • Good grip when handling wet parts due to palm profile
  • Flexible in warmth an cold, good dexterity
  • Seamless tricot allows a very fine touch
Ideal for removal of contaminated parts from dipping baths, handling contaminated heavy parts, cleaning and maintenance in chemical plants and when working with oily and greasy parts.
Certificeringer: EN 388:2016 (2121X), EN ISO 374-1 Type A (AJKLMS); Cat. III; AQL < 0,65
Food contact approved.