Heat resistant gloves, Thermoplus® 955
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
955.8 955.10 955.9
KCHL955.8EA 382 DKK
KCHL955.8 KCHL955.10 KCHL955.9
Heat resistant gloves, Thermoplus® 955
Handsker
This heat protective glove is made of several different materials to provide the best possible protection against heat while offering a good mechanical resistance. The glove protects against a contact heat up to 100 °C. The cotton lining provides comfort, while the para-aramide offers additional cut protection.
  • Thermal insulation, cut resistance
  • Excellent fine touch
  • Good sweat absorption for high wearing comfort
  • Additional cut and heat protection thanks to long knit wrist
Ideal for use in metal and plastics processing, automotive industries, laboratories (when working with heat ovens and cabinet dryers) and foundries.
Certificeringer: EN 388:2016 (344XB), EN 407 (X13XXX); Cat. III