Industrial gloves, 380
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
380-L 380-M 380-S 380-XL
112-5821EA 572 DKK
112-5821 112-5822 112-5823 112-5824
Industrial gloves, 380
Handsker
These seamless knitted nylon gloves feature a microporous nitrile coating on the palm which protects the hand and provides an optimal grip.
  • Palm coating
  • Foamed embossed finish for optimal grip in dry and slightly oily environments
  • Elasticated cuff, anatomical shape
  • Lightweight and low-linting
  • Robust, great dexterity and good tearing resistance
  • Breathable back reduces perspiration
Ideal for light assembly tasks in construction industries and in the maritime sector.
Certificeringer: EN 388:2016 (3121X); Cat. II