Glove liners, BO900
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BO900-UNI BO900-XL
112-3483EA 297 DKK
112-3483 112-3484
Glove liners, BO900
Handsker
These seamless knitted gloves made of nylon and polyester are ideal for use as undergloves.
  • Low linting, lightweight
  • Optimal dexterity
  • Exceptional capacity to absorb perspiration
  • Can be washed at 40 °C using a neutral detergent
Suitable for use in the following industries: Electronic components, Quality Control and integrated circuits.
Certificeringer: Cat. I