Chemical resistant gloves, Showa B0710
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B0710-S B0710-M B0710-L B0710-XL B0710CP-L B0710CP-M B0710CP-S B0710CP-XL
112-5993EA 1240 DKK
112-5993 112-5992 112-5991 112-5994 112-5972 112-5973 112-5974 112-5975
Chemical resistant gloves, Showa B0710
Handsker
These unsupported, non powdered PVC gloves protect the hand against chemicals. As they are impermeable they can be used in damp or greasy environments.
  • Anatomical shape, remain flexible during wear
  • Thin, light gloves with a 'second skin' feel
  • Surface enables a secure grip on slippery objects
  • Easy to put on and remove, lint- and dust-free
  • Extended protection for the entire arm
  • Available in 'clean packed' version
Ideal for people working in health environments and petrochemical industries.
Thickness: 0,30 mm
Colour: White
EN 374-2/-3, EN 388 (3000); Cat. III
Certificeringer: EN ISO 374-1 Type C (JK), EN ISO 374-5, EN 388:2016 (2000X); Cat. III