Glove liners, B0610
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BO610-S BO610-M BO610-L BO610-XL
112-5989EA 286 DKK
112-5989 112-5988 112-5987 112-5990
Glove liners, B0610
Handsker
These seamless knitted gloves with elasticated cuffs are made of nylon. They are uncoated and provide optimal dexterity and tactile feeling.
  • Lightweight, elastic, low-linting
  • Resistant to deformation
  • Minimal allergy risk, no irritation by seams
  • Great freedom of movement for the wrist
Ideal for light assembly tasks in logistics or in the metal industry.
Certificeringer: Cat. I