Examination gloves, Peha-soft®
Leverandør: HARTMANN PAUL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9421605 9421615 9421595 9421635
HART9421605EA 155 DKK
HART9421605 HART9421615 HART9421595 HART9421635
Examination gloves, Peha-soft®
Handsker
These disposable examination gloves made of soft latex are ideal for wearers who experience hypersensitivity reactions to glove powder. Also suitable for use in all areas where staff are at particular risk of contamination and need high touch sensitivity and a good fit. May be used in direct contact with food.
  • Non sterile, powder-free
  • Highly elastic and tear-resistant
  • Texturing of the finger area ensures optimum grip
  • Double-chlorination for excellent donning, fit and high tactile sensitivity
Certificeringer: Food contact approved