High-purity tubing, Tygon® 2475
Leverandør: Saint Gobain Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ACG00003 ACG00007 ACG00012 ACG00017 ACG00022 ACG00027 ACG00038 ACG00046 ACG00053 ACG42064
VERNACG00003EA 1190 DKK
VERNACG00003 VERNACG00007 VERNACG00012 VERNACG00017 VERNACG00022 VERNACG00027 VERNACG00038 VERNACG00046 VERNACG00053 VERNACG42064
High-purity tubing, Tygon® 2475
Slanger Overførselsslange
Tygon®, High-purity tubing providing a high level of purity not previously available in a clear, flexible tubing.
  • Exceptionally low absorption and adsorption compared to silicone
  • Smooth inner surface inhibits particulate entrapment
  • Environmentally safer to dispose
  • Highly resistant to aggressive cleaners and sanitisers
  • Plasticiser free
Applications includes sterile filling and dispensing systems, diagnostic and nuclear equipment, laboratory analytical instrumentation, infusion sets for parenterals and drugs, cosmetic production, food and beverage processing, cell and tissue culture transport.
Temperature resistance: –70 to +52 °C
Hardness (Shore durometer A): 72
Density: 0,90
Certificeringer: Meets USP class VI and FDA criteria.