Diluent for western blotting, Swift™ Western Diluent
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-158 786-679
786-158EA 4210 DKK
786-158 786-679
Diluent for western blotting, Swift™ Western Diluent
Elektroforese reagenser Western Blotting-reagenser
The Swift™ Western Diluent is a solution that reduces the blocking and antibody incubations on Western blot membranes to <60 minutes. Swift™ Western Diluent generates comparable results to traditional Western blotting procedures and other commercial “fast” Western blotting kits.
  • Affordable: Single reagent as opposed to kit or equipment-based
  • Fast: Reduce blot development to <60 mins
  • Versatile: Compatible with all combinations of primary and secondary antibodies
  • For all wet, semi-dry and automated blotting systems
For added convenience, the SWIFT™ Western Diluent is also supplied as a complete kit to ensure optimal results. The SWIFT™ Western Blotting System kit includes SWIFT™ Western Diluent, proprietary wash buffers and femtoLUCENT™ chemiluminescence detection reagent.