Cell culture flasks, Nunclon™ Sphera™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
174951 174952
392-0379EA 2410 DKK
392-0379 392-0380
Cell culture flasks, Nunclon™ Sphera™
Kolber Celledyrkningskolber
The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids. Studies show that over time the spheroids grow in volume, indicating that the new surface enables consistent, repeatable cell growth.
  • Optimises spheroid formation
  • Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
  • Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimises variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth, providing a better model system for studying tumour cell progression and efficacy of anticancer agents in vitro.
As an important milestone in the differentiation process, the Nunclon™ Sphera™ surface demonstrates superior quality for embryoid body (EB) formation of pluripotent stem cells with minimal spontaneous differentiation. The resultant cells are able to differentiate into all three germ layers.