Cleanroom dispensers for personal protective equipment
Leverandør: Palbam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GD-8060 GD-6060 GD-8540 GD-4060 GD-2060
134-0504EA 15790 DKK
134-0504 134-0503 134-0502 113-7992 113-8052
Cleanroom dispensers for personal protective equipment
PPE-dispensere
These wall-mounted dispensers are designed for clean and efficient dispensing and storage.
  • No longer particles build-up
  • Stainless steel 304 electropolished finish
  • Smooth curved opening for easy and snag-free pull out
Suitable for gloves, hairnets, face masks and over shoes in the cleanroom industries, such as pharmaceutical, biotechology, medical device, microelectronics and nanotechnology.
Certificeringer: ISO certified
Leveringsinformation: Typically available from stock for direct shipment from the supplier.