Cleanroom tapes, CR100PC™
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CR100PC-1LB CR100PC-2LB CR100PC-12DB CR100PC-1DB CR100PC-2DB CR100PC-14WH CR100PC-12WH CR100PC-2WH CR100PC-14YE CR100PC-1YE CR100PC-2YE CR100PC-12DG CR100PC-2DG CR100PC-14RE CR100PC-12RE CR100PC-2RE CR100PC-12TR CR100PC-1TR CR100PC-2TR CR100PC-1WH CR100PC-3DB CR100PC-15DB_U CR100PC-38TR CR100PC-38DB CR100PC-38RE CR100PC-38YE CR100PC-3DG CR100PC-3RE CR100PC-3WH CR100PC-4TR CR100PC-38DG CR100PC-15TR CR100PC-15WH CR100PC-15YE CR100PC-34WH CR100PC-15LB CR100PC-2LG CR100PC-1LG CR100PC-38WH CR100PC-4WH CR100PC-12WHPK CR100PC-15DB CR100PC-1DG CR100PC-1OR CR100PC-1RE CR100-34TRIRM CR100-12WHIRM
MICRCR100PC-1LBEA 8590 DKK
MICRCR100PC-1LB MICRCR100PC-2LB 811-0205 MICRCR100PC-1DB MICRCR100PC-2DB MICRCR100PC-14WH MICRCR100PC-12WH MICRCR100PC-2WH MICRCR100PC-14YE MICRCR100PC-1YE MICRCR100PC-2YE MICRCR100PC-12DG MICRCR100PC-2DG MICRCR100PC-14RE MICRCR100PC-12RE MICRCR100PC-2RE MICRCR100PC-12TR MICRCR100PC-1TR MICRCR100PC-2TR 819-0020 MICRCR100PC-3DB MICRCR100PC-15DB_U MICRCR100PC-38TR MICRCR100PC-38DB 811-0206 MICRCR100PC-38YE MICRCR100PC-3DG MICRCR100PC-3RE MICRCR100PC-3WH MICRCR100PC-4TR MICRCR100PC-38DG MICRCR100PC-15TR MICRCR100PC-15WH MICRCR100PC-15YE MICRCR100PC-34WH MICRCR100PC-15LB MICRCR100PC-2LG MICRCR100PC-1LG MICRCR100PC-38WH MICRCR100PC-4WH MICRCR100PC-12WHPK MICRCR100PC-15DB MICRCR100PC-1DG MICRCR100PC-1OR MICRCR100PC-1RE MICRCR100-34TRIRM MICRCR100-12WHIRM
Cleanroom tapes, CR100PC™
Tape
Extremely conformable, vinyl cleanroom tape with synthetic rubber, clean-peel adhesive; ideal for colour coding, sealing and marking on curved and irregular surfaces in cleanroom maintenance and support areas.
  • Clean peel from wafer cartridges and cleanroom work surfaces
  • Synthetic rubber adhesive ensures a clean peel from most cleanroom surfaces such as polyethylene, plastics and stainless steel
  • Matte finish can be written on with a cleanroom pen
  • Ideal for taping and sealing curved and irregular surfaces
  • Special processing and packaging techniques ensure low particles and extractable ion counts for applications in and around the cleanroom
  • Wound on a 76 mm (3") plastic core
Length: 33 m
Available in a variety of colours and widths.
Certificeringer: Complies with Federal Specification PPP-T-66E and CID A-A 1689A.