Nunc™ Coloured Closures for CryoTube™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
363436 336078 336088 336081 336092 336094
391-7638EA 3001 DKK
391-7638 479-2476 479-2478 479-2477 479-2479 479-2480
Nunc™ Coloured Closures for CryoTube™
Låg Låg til kryorør
Colour-code your sample vials. Thermo Scientific™ Nunc™ CryoTube™ Coloured caps are for use with Nunc internally threaded CryoTube vials and include a silicone gasket to provide the best seal. Nunc CryoTubes are the most widely used cryo vials in the world, stemming from the first cryo vials manufactured in the early 1960s. The well-proven design, the high certification level and the large portfolio of vials provide researchers with a choice of the vial best suited for their application and peace of mind that the sample can be retrieved viable and unharmed.
  • These replacement polypropylene screw caps have a silicone gasket to provide the best seal
  • Available nonsterile or irradiated