ChromBox kit
Leverandør: VICI JOUR RESEARCH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JR-802 JR-804 JR-806 JR-357180 JR-357090 JR-34N JR-9001 JR-LZN1-5
JOURJR-802EA 683 DKK
JOURJR-802 JOURJR-804 JOURJR-806 JOURJR-357180 JOURJR-357090 JOURJR-34N JOURJR-9001 JOURJR-LZN1-5
ChromBox kit
Kromatografi forsyninger Kromatografi reservedele
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
Bestillingsoplysninger: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.