Leather gloves, Goodjob® Lahore
Leverandør: FELDTMANN HELMUT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0274/9 0274/8 0274/7 0274/10
112-2972EA 0 DKK
112-2972 FELD0274/8 FELD0274/7 FELD0274/10
Leather gloves, Goodjob® Lahore
Handsker
These gloves without cuff are made of sheep nappa leather.
  • With breathable tricot cotton back
  • Soft leather
  • Good sense of touch
Ideal for general applications in the automobile industry, mechanical engineering, aerospace industry and in the plastic industry.
EN 388 (1111)
Certificeringer: EN 388:2016 (1101X)