Cell culture media, Medium 199
Leverandør: Biowest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L0356-500 L0330-500 L0355-500 P0410-N1L P0410-N10L P0420-N1L P0420-N10L P0425-N1L P0425-N10L L0361-500 P0420-N50L P0425-N50L P0410-N50L P0420-N5L
L0356-500EA 133 DKK
L0356-500 L0330-500 L0355-500 P0410-N1L P0410-N10L P0420-N1L P0420-N10L P0425-N1L P0425-N10L L0361-500 P0420-N50L P0425-N50L P0410-N50L P0420-N5L
Cell culture media, Medium 199
Celledyrkningsmedier
The long term cultivation of cells requires the addition of serum. When properly supplemented, Medium 199 has broad species applicability, particularly for cultivation of non-transformed cells. It is widely used in virology, vaccine production and in vitro cultivation of primary explants of mouse pancreatic epithelial and rat lens tissues.