Cell culture media, Ham's F10 (F-10 Nutrient Medium)
Leverandør: Biowest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L0140-500 L0145-500 P0146-N1L P0146-N10L L0130-500 P0146-N50L P0146-N5L
L0140-500EA 82.4 DKK
L0140-500 L0145-500 P0146-N1L P0146-N10L L0130-500 P0146-N50L P0146-N5L
Cell culture media, Ham's F10 (F-10 Nutrient Medium)
Celledyrkningsmedier
Ham's F10 medium is intended to support the growth of Chinese Hamster Ovary cells under serum-free conditions and other mammalian cell types with serum supplementation. It is a popular medium for growth of fastidious cell lines.