Instrument box, anodised aluminium
Leverandør: Agencinox
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9030 9036
AGEN9030EA 402 DKK
AGEN9030 AGEN9036
Instrument box, anodised aluminium
Bakker/Tallerkener/Potter Instrumentbakker/-skåle
Anodised aluminium perforated box.
  • Excellent temperature resistance
  • Good chemical resistance
  • Autoclavable up to 180 °C