Microtube racks, 96-well, reversible, with lid
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M-021AM M-021V M-021R
212-8911EA 1420 DKK
212-8911 212-8912 212-8913
Microtube racks, 96-well, reversible, with lid
Stativer Tube Racks
PP, reversible base with lid.
  • Clear polypropylene lid for better sample viewing
  • Both sides are alphanumerically labelled for better sample location
  • Frosted writing patches on all four sides for smear resistant labelling
Reversible rack designed with wide spacing to store up to 96 microtubes on either side. One side accommodates 1,5/2,0 ml microtubes and the reverse side is for 0,5 ml tubes.