Microcentrifuge tubes
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
409110.1 409110.4 409110.3 409113.2 409113.4
DELT409110.1EA 1490 DKK
DELT409110.1 DELT409110.4 DELT409110.3 DELT409113.2 DELT409113.4
Microcentrifuge tubes
Rør Mikrocentrifugerør
Made of PP, transparent.
  • Autoclavable
  • Temperature resistance: −190 to +121 °C
  • Certified RNAse, DNAse and pyrogenfree
  • Screw caps supplied seperately
  • Tubes and caps can be centrifuged up to 17000×g