Safety ankle boots, lace-up, Bacou Ballast
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6245564/38 6245564/39 6245564/40 6245564/41 6245564/42 6245564/43 6245564/44 6245564/45 6245564/46 6245564/47
DALL6245564/38EA 1880 DKK
DALL6245564/38 DALL6245564/39 DALL6245564/40 DALL6245564/41 DALL6245564/42 DALL6245564/43 DALL6245564/44 DALL6245564/45 DALL6245564/46 DALL6245564/47
Safety ankle boots, lace-up, Bacou Ballast
Sko og støvler
Safety ankle boot made from water resistant leather with reflective textile inserts, Coolmax® lining and dual-density PU sole.
  • Padded ankle collar and tongue plus reinforced over cap
  • Antibacterial, quick drying Coolmax® lining
  • Removable Coolmax® insole antibacterial, carbon-treated
  • Light, metal-free Spacium™ toecap in injected polymer (200 J)
  • FLEXIUM ™ metal-free, puncture resistant midsole
For environments like frozen ground or very high temperatures.
Colour: Black
Certificeringer: EN ISO 20345; S3, SRC, CI, HI